Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LTK
Entrez Gene ID
4058
Gene Name
leukocyte receptor tyrosine kinase
Gene Aliases
TYK1
Chromosome Location
Chr.15: 41503639 - 41513887 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135685.1 NP_001129157.1 5 - 6 838 142
NM_002344.5 NP_002335.2 5 - 6 838 142
NM_206961.3 NP_996844.1 5 - 6 838 142
XM_011521556.2 XP_011519858.1 5 - 6 838 142
XM_011521557.2 XP_011519859.1 5 - 6 838 142
XM_017022181.1 XP_016877670.1 5 - 6 838 142
XM_017022182.1 XP_016877671.1 5 - 6 838 142
XM_017022183.1 XP_016877672.1 5 - 6 838 142
GenBank mRNA AK296892.1 - 5 - 6 838 142
AK314006.1 - 5 - 6 805 142
BC045607.1 - 5 - 6 811 142
D16105.1 - 5 - 6 757 142

View Full Product Details