The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136040.2 | NP_001129512.1 | 1 - 2 | 78 | 78 |
NM_001142565.1 | NP_001136037.1 | 1 - 2 | 78 | 78 | |
XM_011545260.2 | XP_011543562.1 | 1 - 2 | 78 | 78 | |
GenBank mRNA | AL832667.1 | - | 1 - 2 | 69 | 78 |
BX537888.1 | - | 1 - 2 | 59 | 78 | |
DA804579.1 | - | 1 - 2 | 78 | 78 |