The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319952.1 | NP_001306881.1 | 13 - 14 | 1415 | 60 |
NM_024526.3 | NP_078802.2 | 14 - 15 | 1467 | 60 | |
XM_011542135.2 | XP_011540437.1 | 14 - 15 | 1509 | 60 | |
XM_017002328.1 | XP_016857817.1 | 14 - 15 | 1392 | 60 | |
XM_017002329.1 | XP_016857818.1 | 14 - 15 | 1368 | 60 | |
GenBank mRNA | AK025175.1 | - | 14 - 15 | 1369 | 60 |
AK225235.1 | - | 14 - 15 | 1369 | 60 | |
AK290261.1 | - | 14 - 15 | 1394 | 60 | |
AK307932.1 | - | 13 - 14 | 1313 | 60 | |
AK310979.1 | - | 14 - 15 | 1454 | 60 |