The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005455.4 | NP_005446.2 | 3 - 4 | 513 | 71 |
NM_203350.2 | NP_976225.1 | 3 - 4 | 513 | 71 | |
GenBank mRNA | AB209568.1 | - | 3 - 4 | 226 | 71 |
AF065391.1 | - | 3 - 4 | 253 | 71 | |
AF065392.1 | - | 3 - 4 | 253 | 71 | |
AK222858.1 | - | 3 - 4 | 238 | 71 | |
AL136945.1 | - | 3 - 4 | 235 | 71 | |
BC033648.1 | - | 3 - 4 | 259 | 71 | |
BC039814.1 | - | 3 - 4 | 259 | 71 | |
DA523601.1 | - | 3 - 4 | 513 | 71 |