The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002395.5 | NP_002386.1 | 11 - 12 | 1536 | 102 |
XM_011535836.2 | XP_011534138.1 | 10 - 11 | 1261 | 102 | |
GenBank mRNA | AK223417.1 | - | 11 - 12 | 1362 | 102 |
AK289783.1 | - | 11 - 12 | 1364 | 102 | |
AK301875.1 | - | 9 - 10 | 1078 | 102 | |
AK302777.1 | - | 10 - 11 | 1261 | 102 | |
BC017403.1 | - | 2 - 3 | 256 | 102 | |
BC025246.1 | - | 11 - 12 | 1352 | 102 | |
X77244.1 | - | 11 - 12 | 1271 | 102 |