The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002395.5 | NP_002386.1 | 4 - 5 | 700 | 83 |
XM_011535836.2 | XP_011534138.1 | 3 - 4 | 425 | 83 | |
GenBank mRNA | AK223417.1 | - | 4 - 5 | 526 | 83 |
AK289783.1 | - | 4 - 5 | 528 | 83 | |
AK301875.1 | - | 2 - 3 | 242 | 83 | |
AK302777.1 | - | 3 - 4 | 425 | 83 | |
BC025246.1 | - | 4 - 5 | 516 | 83 | |
BX376125.2 | - | 4 - 5 | 540 | 83 | |
L34035.1 | - | 4 - 5 | 447 | 83 | |
U43944.1 | - | 4 - 5 | 414 | 83 | |
X77244.1 | - | 4 - 5 | 435 | 83 |