The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282913.1 | NP_001269842.1 | 5 - 6 | 819 | 150 |
NM_001282914.1 | NP_001269843.1 | 5 - 6 | 925 | 150 | |
NM_001282915.1 | NP_001269844.1 | 5 - 6 | 743 | 150 | |
NM_173826.3 | NP_776187.2 | 5 - 6 | 1001 | 150 | |
XM_017006251.1 | XP_016861740.1 | 6 - 7 | 1953 | 150 | |
GenBank mRNA | AK124087.1 | - | 2 - 3 | 240 | 150 |
AK292756.1 | - | 5 - 6 | 819 | 150 | |
AL832473.1 | - | 5 - 6 | 678 | 150 | |
AL833118.1 | - | 3 - 4 | 415 | 150 | |
BC047231.1 | - | 5 - 6 | 620 | 150 | |
BC101059.2 | - | 5 - 6 | 602 | 150 | |
BC101060.2 | - | 5 - 6 | 678 | 150 | |
BC101061.2 | - | 5 - 6 | 602 | 150 |