The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005458.7 | NP_005449.5 | 9 - 10 | 1842 | 70 |
XM_005252316.4 | XP_005252373.1 | 7 - 8 | 875 | 70 | |
XM_017015331.1 | XP_016870820.1 | 8 - 9 | 1260 | 70 | |
XM_017015332.1 | XP_016870821.1 | 6 - 7 | 829 | 70 | |
GenBank mRNA | AF056085.1 | - | 9 - 10 | 1839 | 70 |
AF069755.1 | - | 9 - 10 | 1538 | 70 | |
AF074483.1 | - | 9 - 10 | 1382 | 70 | |
AF099033.1 | - | 9 - 10 | 1382 | 70 | |
AJ012188.1 | - | 9 - 10 | 1382 | 70 | |
AK308139.1 | - | 8 - 9 | 1015 | 70 | |
BC035071.2 | - | 10 - 11 | 1677 | 70 | |
JQ308173.1 | - | 9 - 10 | 1382 | 70 |