Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NEK11
- Entrez Gene ID
-
79858
- Gene Name
- NIMA related kinase 11
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 131026850 - 131350465 on Build GRCh38
View Full Product Details