The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000487.5 | NP_000478.3 | 1 - 2 | 622 | 96 |
NM_001085425.2 | NP_001078894.2 | 2 - 3 | 517 | 96 | |
NM_001085426.2 | NP_001078895.2 | 2 - 3 | 469 | 96 | |
NM_001085427.2 | NP_001078896.2 | 2 - 3 | 424 | 96 | |
XM_011530691.2 | XP_011528993.1 | 1 - 2 | 600 | 96 | |
XM_017028800.1 | XP_016884289.1 | 2 - 3 | 441 | 96 | |
GenBank mRNA | AB448736.1 | - | 1 - 2 | 596 | 96 |
AK092752.1 | - | 2 - 3 | 447 | 96 | |
AK301098.1 | - | 1 - 2 | 619 | 96 | |
AK310564.1 | - | - | 977 | 96 | |
AK315011.1 | - | 2 - 3 | 468 | 96 | |
BC014210.2 | - | 1 - 2 | 592 | 96 | |
BI770997.1 | - | 2 - 3 | 499 | 96 | |
BP306351.1 | - | 2 - 3 | 266 | 96 | |
BX648618.1 | - | 2 - 3 | 438 | 96 | |
CN409235.1 | - | 2 - 3 | 447 | 96 | |
CR456383.1 | - | 1 - 2 | 292 | 96 | |
KU177918.1 | - | 1 - 2 | 222 | 96 | |
KU177919.1 | - | 1 - 2 | 222 | 96 | |
X52151.1 | - | 1 - 2 | 580 | 96 |