The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287821.1 | NP_001274750.1 | 10 - 11 | 1320 | 90 |
NM_001287822.1 | NP_001274751.1 | 9 - 10 | 1224 | 90 | |
NM_001287823.1 | NP_001274752.1 | 9 - 10 | 1173 | 90 | |
NM_017975.4 | NP_060445.3 | 10 - 11 | 1225 | 90 | |
GenBank mRNA | AK000898.1 | - | 8 - 9 | 829 | 90 |
AK023175.1 | - | 10 - 11 | 1215 | 90 | |
AK122787.1 | - | 9 - 10 | 1012 | 90 | |
AK291048.1 | - | 9 - 10 | 930 | 90 | |
BC036900.1 | - | 10 - 11 | 971 | 90 | |
BC090041.1 | - | 10 - 11 | 1069 | 90 | |
BX640701.1 | - | 10 - 11 | 1287 | 90 |