Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QPRT
Entrez Gene ID
23475
Gene Name
quinolinate phosphoribosyltransferase
Gene Aliases
HEL-S-90n, QPRTase
Chromosome Location
Chr.16: 29670588 - 29698699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005255223.3 XP_005255280.2 1 - 2 199 97
XM_017023101.1 XP_016878590.1 1 - 2 199 97

View Full Product Details