Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FECH
Entrez Gene ID
2235
Gene Name
ferrochelatase
Gene Aliases
EPP, FCE
Chromosome Location
Chr.18: 57544841 - 57586737 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000140.3 NP_000131.2 5 - 6 716 91
NM_001012515.2 NP_001012533.1 5 - 6 734 91
XM_011525881.1 XP_011524183.1 5 - 6 705 91
XM_011525882.2 XP_011524184.1 5 - 6 699 91
XM_017025614.1 XP_016881103.1 5 - 6 687 91
GenBank mRNA AK092416.1 - 5 - 6 651 91
AK223190.1 - 5 - 6 591 91
AK292937.1 - 5 - 6 600 91
AK299643.1 - 5 - 6 688 91
BC039841.1 - 5 - 6 683 91
BT019959.1 - 5 - 6 599 91
BX571744.1 - 5 - 6 595 91
D00726.1 - 5 - 6 628 91

View Full Product Details