Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOG
Entrez Gene ID
4340
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
BTN6, BTNL11, MOGIG2, NRCLP7
Chromosome Location
Chr.6: 29656981 - 29672372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170418.1 NP_001163889.1 1 - 2 317 84
NM_206814.5 NP_996537.3 1 - 2 317 84
GenBank mRNA AK308949.1 - 1 - 2 295 84
AY566846.1 - 1 - 2 82 84
AY566847.1 - 1 - 2 82 84
U18800.2 - 1 - 2 88 84

View Full Product Details