Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MOG
Entrez Gene ID
4340
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
BTN6, BTNL11, MOGIG2, NRCLP7
Chromosome Location
Chr.6: 29656981 - 29672372 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002433.4 NP_002424.3 4 - 5 806 95
NM_206809.3 NP_996532.2 4 - 5 806 95
NM_206814.5 NP_996537.3 3 - 4 458 95
XM_005249131.3 XP_005249188.1 4 - 5 795 95
GenBank mRNA AB209815.1 - 4 - 5 716 95
AK295213.1 - 5 - 6 627 95
AK308949.1 - 3 - 4 436 95
AK312892.1 - 4 - 5 692 95
AY566852.1 - 4 - 5 577 95
BC035938.1 - 4 - 5 695 95
U18798.1 - 4 - 5 577 95
U18800.2 - 3 - 4 229 95
U18801.1 - 4 - 5 577 95
U64564.1 - 4 - 5 739 95
U64565.1 - 3 - 4 490 95
U64570.1 - 4 - 5 628 95
U64571.1 - 4 - 5 628 95
X74511.1 - 4 - 5 577 95

View Full Product Details