The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018008.1 | NP_001018008.1 | 1 - 2 | 271 | 65 |
NM_001301289.1 | NP_001288218.1 | 1 - 2 | 271 | 65 | |
XM_005254646.2 | XP_005254703.1 | 1 - 2 | 257 | 65 | |
XM_005254647.3 | XP_005254704.1 | 1 - 2 | 257 | 65 | |
XM_005254650.3 | XP_005254707.1 | 1 - 2 | 257 | 65 | |
XM_005254651.1 | XP_005254708.1 | 1 - 2 | 257 | 65 | |
XM_005254652.1 | XP_005254709.1 | 1 - 2 | 257 | 65 | |
XM_005254653.2 | XP_005254710.1 | 1 - 2 | 257 | 65 | |
XM_017022540.1 | XP_016878029.1 | 1 - 2 | 257 | 65 | |
XM_017022541.1 | XP_016878030.1 | 1 - 2 | 257 | 65 | |
GenBank mRNA | AB209041.1 | - | 1 - 2 | 308 | 65 |
AK298625.1 | - | 1 - 2 | 147 | 65 | |
AK301319.1 | - | 1 - 2 | 167 | 65 | |
BC050473.1 | - | 1 - 2 | 258 | 65 | |
BC053545.1 | - | 1 - 2 | 269 | 65 | |
DQ424903.1 | - | 1 - 2 | 132 | 65 |