Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD8A
Entrez Gene ID
925
Gene Name
CD8a molecule
Gene Aliases
CD8, Leu2, MAL, p32
Chromosome Location
Chr.2: 86784605 - 86808396 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145873.1 NP_001139345.1 7 - 8 1654 65
NM_001768.6 NP_001759.3 4 - 5 1512 65
NR_027353.1 5 - 6 1098 65
GenBank mRNA AK097942.1 - 5 - 6 697 65
AK124156.1 - 7 - 8 1654 65
AK300089.1 - 5 - 6 1064 65
AY039664.1 - 4 - 5 802 65
BC025715.1 - 4 - 5 628 65
M12824.1 - 4 - 5 710 65
M12828.1 - 4 - 5 688 65

View Full Product Details