The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015719.3 | NP_056534.2 | 4 - 5 | 677 | 109 |
XM_011528042.2 | XP_011526344.1 | 4 - 5 | 735 | 109 | |
GenBank mRNA | AB209175.1 | - | 2 - 3 | 330 | 109 |
AF177941.1 | - | 4 - 5 | 677 | 109 |