The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019098.4 | NP_061971.3 | 10 - 11 | 1219 | 90 |
XM_011517138.2 | XP_011515440.1 | 8 - 9 | 874 | 90 | |
GenBank mRNA | AF228520.1 | - | 7 - 8 | 832 | 90 |
AF272900.1 | - | 10 - 11 | 1217 | 90 | |
BC150601.1 | - | 10 - 11 | 1197 | 90 |