The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019098.4 | NP_061971.3 | 12 - 13 | 1523 | 85 |
XM_011517138.2 | XP_011515440.1 | 10 - 11 | 1178 | 85 | |
GenBank mRNA | AF228520.1 | - | 9 - 10 | 1136 | 85 |
AF272900.1 | - | 12 - 13 | 1521 | 85 | |
BC150601.1 | - | 12 - 13 | 1501 | 85 |