The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019098.4 | NP_061971.3 | 14 - 15 | 1716 | 99 |
XM_011517138.2 | XP_011515440.1 | 12 - 13 | 1371 | 99 | |
GenBank mRNA | AF228520.1 | - | 11 - 12 | 1329 | 99 |
AF272900.1 | - | 14 - 15 | 1714 | 99 | |
BC150601.1 | - | 14 - 15 | 1694 | 99 |