Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
Chromosome Location
Chr.8: 86573608 - 86743675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019098.4 NP_061971.3 1 - 2 176 82
GenBank mRNA AF272900.1 - 1 - 2 174 82
AL713036.1 - 1 - 2 177 82
BC150601.1 - 1 - 2 154 82

View Full Product Details