The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019098.4 | NP_061971.3 | 9 - 10 | 1106 | 106 |
XM_011517138.2 | XP_011515440.1 | 7 - 8 | 761 | 106 | |
GenBank mRNA | AF228520.1 | - | 6 - 7 | 719 | 106 |
AF272900.1 | - | 9 - 10 | 1104 | 106 | |
BC150601.1 | - | 9 - 10 | 1084 | 106 |