Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37C
Entrez Gene ID
55048
Gene Name
VPS37C, ESCRT-I subunit
Gene Aliases
-
Chromosome Location
Chr.11: 61130256 - 61161444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK302384.1 - 1 - 2 121 182

View Full Product Details