The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159286.1 | NP_001152758.1 | 14 - 15 | 1833 | 116 |
NM_032019.5 | NP_114408.3 | 15 - 16 | 1893 | 116 | |
GenBank mRNA | AF393962.1 | - | 15 - 16 | 1520 | 116 |
AF407272.1 | - | 15 - 16 | 1520 | 116 | |
AF407273.1 | - | 14 - 15 | 1460 | 116 | |
AF426160.1 | - | 15 - 16 | 1873 | 116 | |
AY450395.1 | - | 12 - 13 | 1285 | 116 | |
BC032049.1 | - | 15 - 16 | 1706 | 116 | |
BC094734.1 | - | 13 - 14 | 1561 | 116 | |
BC125082.1 | - | 9 - 10 | 1042 | 116 | |
BC125083.1 | - | 15 - 16 | 1567 | 116 | |
CR456465.1 | - | 15 - 16 | 1576 | 116 |