Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A16
Entrez Gene ID
85413
Gene Name
solute carrier family 22 member 16
Gene Aliases
CT2, FLIPT2, HEL-S-18, OAT6, OCT6, OKB1, dJ261K5.1
Chromosome Location
Chr.6: 110424587 - 110476641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011536209.2 XP_011534511.1 2 - 3 240 99

View Full Product Details