Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB48
Entrez Gene ID
3104
Gene Name
zinc finger and BTB domain containing 48
Gene Aliases
HKR3, ZNF855, pp9964
Chromosome Location
Chr.1: 6579967 - 6589280 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278647.1 NP_001265576.1 1 - 2 56 72
GenBank mRNA AK290156.1 - 1 - 2 51 72

View Full Product Details