The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080401.1 | NP_001073870.1 | 2 - 3 | 1058 | 109 |
XM_011526474.2 | XP_011524776.1 | 2 - 3 | 1567 | 109 | |
GenBank mRNA | AK097444.1 | - | 1 - 2 | 1461 | 109 |
BC062452.1 | - | 2 - 3 | 236 | 109 |