Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF4A3
Entrez Gene ID
9775
Gene Name
eukaryotic translation initiation factor 4A3
Gene Aliases
DDX48, MUK34, NMP265, NUK34, RCPS, eIF4AIII
Chromosome Location
Chr.17: 80135214 - 80147183 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014740.3 NP_055555.1 11 - 12 1445 79
XM_011525522.1 XP_011523824.1 10 - 11 1338 79
GenBank mRNA AK290608.1 - 11 - 12 1426 79
BC003662.2 - 11 - 12 1355 79
BC004386.1 - 11 - 12 1376 79
BC011151.1 - 11 - 12 1380 79
CA428254.1 - 1 - 2 79
D21853.1 - 11 - 12 1437 79
X79538.1 - 11 - 12 1296 79

View Full Product Details