The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243342.1 | NP_001230271.1 | 11 - 12 | 1839 | 152 |
NM_017950.3 | NP_060420.2 | 11 - 12 | 1839 | 152 | |
XM_011524963.2 | XP_011523265.1 | 11 - 12 | 1876 | 152 | |
XM_011524964.2 | XP_011523266.1 | 5 - 6 | 808 | 152 | |
XM_011524965.2 | XP_011523267.1 | 11 - 12 | 1839 | 152 | |
XM_017024807.1 | XP_016880296.1 | 11 - 12 | 1839 | 152 | |
GenBank mRNA | AB046860.1 | - | 7 - 8 | 1345 | 152 |
BC035251.1 | - | 11 - 12 | 1832 | 152 |