The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006445.3 | NP_006436.3 | 1 - 2 | 106 | 58 |
GenBank mRNA | BC000579.2 | - | 1 - 2 | 71 | 58 |
BC064370.1 | - | 1 - 2 | 71 | 58 | |
CD698473.1 | - | 1 - 2 | 128 | 58 |