Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC29
Entrez Gene ID
26231
Gene Name
leucine rich repeat containing 29
Gene Aliases
FBL9, FBXL9
Chromosome Location
Chr.16: 67207139 - 67227062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012163.2 NP_036295.1 1 - 2 185 80
XM_017023127.1 XP_016878616.1 1 - 2 335 80
XM_017023132.1 XP_016878621.1 1 - 2 335 80
XM_017023133.1 XP_016878622.1 1 - 2 335 80
GenBank mRNA BC018785.1 - 1 - 2 185 80

Gene Symbol
LOC105369155
Entrez Gene ID
105369155
Gene Name
uncharacterized LOC105369155
Gene Aliases
-
Chromosome Location
Chr.16: 67210350 - 67226998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011523526.2 XP_011521828.1 2 - 3 80 80

View Full Product Details