The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000123.3 | NP_000114.2 | 11 - 12 | 2964 | 142 |
GenBank mRNA | AF462447.1 | - | 11 - 12 | 2921 | 142 |
AK294708.1 | - | 11 - 12 | 2893 | 142 | |
AK303558.1 | - | 12 - 13 | 2680 | 142 | |
BC031522.2 | - | 11 - 12 | 2950 | 142 | |
BX647399.1 | - | 11 - 12 | 2793 | 142 | |
L20046.1 | - | 11 - 12 | 2586 | 142 | |
LM994821.1 | - | 7 - 8 | 2034 | 142 | |
X69978.1 | - | 11 - 12 | 2735 | 142 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204425.1 | NP_001191354.1 | 19 - 20 | 4022 | 142 |
GenBank mRNA | AB209312.1 | - | 19 - 20 | 3814 | 142 |
LM994822.1 | - | 19 - 20 | 3900 | 142 |