Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTC1
Entrez Gene ID
80169
Gene Name
CTS telomere maintenance complex component 1
Gene Aliases
AAF-132, AAF132, C17orf68, CRMCC, tmp494178
Chromosome Location
Chr.17: 8224821 - 8248095 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025099.5 NP_079375.3 20 - 21 3281 65
XM_006721577.3 XP_006721640.1 19 - 20 3136 65
XM_011524010.2 XP_011522312.1 20 - 21 3160 65
XM_011524011.2 XP_011522313.1 19 - 20 3030 65
GenBank mRNA AK025823.1 - 10 - 11 1424 65
AK091077.1 - 20 - 21 3232 65
AK225375.1 - 10 - 11 1424 65
BC026057.2 - 11 - 12 1422 65
BC110373.1 - 12 - 13 1648 65
BC111783.1 - 20 - 21 3232 65
BX406692.2 - 7 - 8 840 65
DQ451688.1 - 20 - 21 3222 65
FJ790670.1 - 20 - 21 3222 65

View Full Product Details