Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN1
Entrez Gene ID
3998
Gene Name
lectin, mannose binding 1
Gene Aliases
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58
Chromosome Location
Chr.18: 59327823 - 59359276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005570.3 NP_005561.1 4 - 5 577 149
GenBank mRNA AK223208.1 - 4 - 5 555 149
AK225108.1 - 4 - 5 555 149
AK225111.1 - 4 - 5 614 149
AK301244.1 - 4 - 5 566 149
AK312869.1 - 4 - 5 555 149
BC017858.1 - 4 - 5 562 149
BC032330.1 - 4 - 5 560 149
U09716.1 - 4 - 5 565 149
X71661.1 - 4 - 5 566 149

View Full Product Details