Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN1
Entrez Gene ID
3998
Gene Name
lectin, mannose binding 1
Gene Aliases
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58
Chromosome Location
Chr.18: 59327823 - 59359276 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005570.3 NP_005561.1 5 - 6 666 147
GenBank mRNA AK223208.1 - 5 - 6 644 147
AK225108.1 - 5 - 6 644 147
AK225111.1 - 5 - 6 703 147
AK312869.1 - 5 - 6 644 147
BC017858.1 - 5 - 6 651 147
BC032330.1 - 5 - 6 649 147
U09716.1 - 5 - 6 654 147
X71661.1 - 5 - 6 655 147

View Full Product Details