The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080480.2 | NP_001073949.1 | 6 - 7 | 758 | 103 |
NR_073465.1 | 3 - 4 | 478 | 103 | ||
XM_006714999.2 | XP_006715062.1 | 6 - 7 | 495 | 103 | |
XM_006715000.3 | XP_006715063.1 | 6 - 7 | 696 | 103 | |
XM_011514313.2 | XP_011512615.1 | 6 - 7 | 696 | 103 | |
GenBank mRNA | AB305043.1 | - | 6 - 7 | 528 | 103 |
AK131269.1 | - | 6 - 7 | 721 | 103 | |
AK304748.1 | - | 6 - 7 | 703 | 103 | |
BC150650.1 | - | 6 - 7 | 704 | 103 | |
BC150652.1 | - | 6 - 7 | 704 | 103 |