The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001448.2 | NP_001439.2 | 4 - 5 | 1093 | 94 |
GenBank mRNA | AF030186.1 | - | 4 - 5 | 1093 | 94 |
AF064826.1 | - | 4 - 5 | 998 | 94 | |
AK304207.1 | - | 4 - 5 | 818 | 94 | |
AK312605.1 | - | 4 - 5 | 1350 | 94 | |
AY052833.1 | - | 4 - 5 | 1017 | 94 | |
AY358507.1 | - | 4 - 5 | 1379 | 94 | |
BC017166.1 | - | 4 - 5 | 1387 | 94 |