The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001448.2 | NP_001439.2 | 8 - 9 | 1669 | 77 |
GenBank mRNA | AF030186.1 | - | 8 - 9 | 1669 | 77 |
AF064826.1 | - | 8 - 9 | 1574 | 77 | |
AK304207.1 | - | 8 - 9 | 1394 | 77 | |
AK312605.1 | - | 8 - 9 | 1926 | 77 | |
AY052833.1 | - | 8 - 9 | 1593 | 77 | |
AY358507.1 | - | 8 - 9 | 1955 | 77 | |
BC017166.1 | - | 8 - 9 | 1963 | 77 |