The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001830.3 | NP_001821.2 | 2 - 3 | 421 | 92 |
GenBank mRNA | AB019432.1 | - | 1 - 2 | 266 | 92 |
AK225287.1 | - | 2 - 3 | 382 | 92 | |
AK289564.1 | - | 2 - 3 | 368 | 92 | |
BC130278.1 | - | 1 - 2 | 280 | 92 | |
BP310547.1 | - | 2 - 3 | 421 | 92 | |
X77197.1 | - | 2 - 3 | 374 | 92 |