The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143943.1 | NP_001137415.1 | 1 - 2 | 107 | 134 |
NM_032328.3 | NP_115704.1 | 2 - 3 | 282 | 134 | |
GenBank mRNA | AK302883.1 | - | 1 - 2 | 50 | 134 |
BC005357.1 | - | 1 - 2 | 46 | 134 | |
BQ438351.1 | - | 1 - 2 | 49 | 134 | |
DB286332.1 | - | 1 - 2 | 109 | 134 |