The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003679.4 | NP_003670.2 | 5 - 6 | 116 | |
XM_005273337.1 | XP_005273394.1 | 5 - 6 | 116 | ||
XM_005273338.3 | XP_005273395.1 | 6 - 7 | 116 | ||
XM_017002649.1 | XP_016858138.1 | 6 - 7 | 116 | ||
XM_017002650.1 | XP_016858139.1 | 3 - 4 | 116 | ||
GenBank mRNA | AF056032.1 | - | 5 - 6 | 116 | |
AK291558.1 | - | 5 - 6 | 428 | 116 | |
AK310937.1 | - | 5 - 6 | 116 | ||
BC005297.1 | - | 5 - 6 | 116 | ||
BP276365.1 | - | 5 - 6 | 116 | ||
Y13153.1 | - | 5 - 6 | 116 |