The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024649.1 | NP_001019820.1 | 13 - 14 | 1772 | 129 |
NM_001746.3 | NP_001737.1 | 13 - 14 | 1825 | 129 | |
XM_011534664.1 | XP_011532966.1 | 13 - 14 | 2190 | 129 | |
XM_011534665.2 | XP_011532967.1 | 13 - 14 | 1798 | 129 | |
GenBank mRNA | AJ271880.1 | - | 12 - 13 | 1664 | 129 |
AK130080.1 | - | 3 - 4 | 307 | 129 | |
AK294702.1 | - | 13 - 14 | 1828 | 129 | |
AK304420.1 | - | 12 - 13 | 1707 | 129 | |
AK312334.1 | - | 13 - 14 | 1732 | 129 | |
BC003552.1 | - | 13 - 14 | 1759 | 129 | |
BC042843.1 | - | 13 - 14 | 1706 | 129 | |
L10284.1 | - | 13 - 14 | 1737 | 129 | |
L18887.1 | - | 13 - 14 | 1743 | 129 | |
M94859.1 | - | 13 - 14 | 1743 | 129 | |
M98452.1 | - | 6 - 7 | 940 | 129 |