The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_046234.1 | 1 - 2 | 166 | 93 | |
GenBank mRNA | AK058090.1 | - | 1 - 2 | 166 | 93 |
AL834142.1 | - | 1 - 2 | 129 | 93 | |
BC036904.1 | - | 1 - 2 | 103 | 93 |