The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320356.1 | NP_001307285.1 | 5 - 6 | 798 | 72 |
NM_001320357.1 | NP_001307286.1 | 4 - 5 | 688 | 72 | |
NM_001320358.1 | NP_001307287.1 | 4 - 5 | 558 | 72 | |
NM_001320359.1 | NP_001307288.1 | 5 - 6 | 714 | 72 | |
NM_001320360.1 | NP_001307289.1 | 4 - 5 | 944 | 72 | |
NM_001320361.1 | NP_001307290.1 | 5 - 6 | 928 | 72 | |
NM_001320362.1 | NP_001307291.1 | 4 - 5 | 508 | 72 | |
NM_001320363.1 | NP_001307292.1 | 4 - 5 | 638 | 72 | |
NM_001320364.1 | NP_001307293.1 | 3 - 4 | 567 | 72 | |
NM_001320365.1 | NP_001307294.1 | 5 - 6 | 878 | 72 | |
NM_015528.2 | NP_056343.1 | 4 - 5 | 1074 | 72 | |
XM_017024427.1 | XP_016879916.1 | 3 - 4 | 814 | 72 | |
GenBank mRNA | AK025329.1 | - | 4 - 5 | 747 | 72 |
AL050060.1 | - | 2 - 3 | 224 | 72 | |
AL521425.3 | - | 5 - 6 | 796 | 72 | |
AL537131.3 | - | 5 - 6 | 602 | 72 | |
AL834284.1 | - | 3 - 4 | 596 | 72 | |
AY203930.1 | - | 3 - 4 | 574 | 72 | |
AY960626.1 | - | 3 - 4 | 241 | 72 | |
BC010139.2 | - | 3 - 4 | 472 | 72 | |
BC013222.1 | - | 3 - 4 | 528 | 72 | |
BI825561.1 | - | 4 - 5 | 410 | 72 | |
CB150714.1 | - | 4 - 5 | 460 | 72 | |
CR457340.1 | - | 3 - 4 | 298 | 72 |