The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320356.1 | NP_001307285.1 | 10 - 11 | 1247 | 82 |
NM_001320357.1 | NP_001307286.1 | 9 - 10 | 1137 | 82 | |
NM_001320358.1 | NP_001307287.1 | 9 - 10 | 1007 | 82 | |
NM_001320359.1 | NP_001307288.1 | 10 - 11 | 1163 | 82 | |
NM_001320360.1 | NP_001307289.1 | 9 - 10 | 1393 | 82 | |
NM_001320361.1 | NP_001307290.1 | 10 - 11 | 1377 | 82 | |
NM_001320362.1 | NP_001307291.1 | 9 - 10 | 957 | 82 | |
NM_001320363.1 | NP_001307292.1 | 9 - 10 | 1087 | 82 | |
NM_001320364.1 | NP_001307293.1 | 8 - 9 | 1016 | 82 | |
NM_001320365.1 | NP_001307294.1 | 10 - 11 | 1327 | 82 | |
NM_015528.2 | NP_056343.1 | 9 - 10 | 1523 | 82 | |
XM_017024427.1 | XP_016879916.1 | 8 - 9 | 1263 | 82 | |
XM_017024428.1 | XP_016879917.1 | 2 - 3 | 548 | 82 | |
GenBank mRNA | AK025329.1 | - | 9 - 10 | 1196 | 82 |
AL050060.1 | - | 7 - 8 | 670 | 82 | |
AL834284.1 | - | 8 - 9 | 1042 | 82 | |
AY203930.1 | - | 8 - 9 | 1023 | 82 | |
BC010139.2 | - | 8 - 9 | 921 | 82 | |
BC013222.1 | - | 8 - 9 | 977 | 82 | |
CR457340.1 | - | 8 - 9 | 747 | 82 |