The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000715.3 | NP_000706.1 | 3 - 4 | 522 | 84 |
XM_005273251.1 | XP_005273308.1 | 3 - 4 | 478 | 84 | |
XM_005273252.4 | XP_005273309.1 | 3 - 4 | 654 | 84 | |
GenBank mRNA | AK303793.1 | - | 4 - 5 | 333 | 84 |
AK313164.1 | - | 3 - 4 | 468 | 84 | |
BC022312.1 | - | 3 - 4 | 522 | 84 | |
M31452.1 | - | 3 - 4 | 466 | 84 | |
X02865.1 | - | 1 - 2 | 91 | 84 |