The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024766.4 | NP_079042.1 | 8 - 9 | 802 | 87 |
XM_011533113.2 | XP_011531415.1 | 6 - 7 | 478 | 87 | |
XM_017004982.1 | XP_016860471.1 | 6 - 7 | 2905 | 87 | |
XM_017004983.1 | XP_016860472.1 | 7 - 8 | 1017 | 87 | |
GenBank mRNA | AK027104.1 | - | 8 - 9 | 742 | 87 |
BC053733.1 | - | 8 - 9 | 729 | 87 |