The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004939.2 | NP_004930.1 | 5 - 6 | 574 | 113 |
GenBank mRNA | AB593031.1 | - | 5 - 6 | 327 | 113 |
AB593032.1 | - | 5 - 6 | 304 | 113 | |
AB593034.1 | - | 5 - 6 | 347 | 113 | |
AK298212.1 | - | 2 - 3 | 158 | 113 | |
AK313933.1 | - | 5 - 6 | 293 | 113 | |
BC012132.1 | - | 5 - 6 | 310 | 113 | |
BC053673.1 | - | 5 - 6 | 311 | 113 | |
EF418570.1 | - | 5 - 6 | 279 | 113 | |
EF418571.1 | - | 5 - 6 | 279 | 113 | |
X70649.1 | - | 5 - 6 | 549 | 113 |