The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048164.2 | NP_001041629.1 | 6 - 7 | 1194 | 83 |
NM_032803.5 | NP_116192.4 | 6 - 7 | 1218 | 83 | |
XM_017029912.1 | XP_016885401.1 | 5 - 6 | 1709 | 83 | |
GenBank mRNA | AF320612.1 | - | 6 - 7 | 1169 | 83 |
AK027447.1 | - | 4 - 5 | 540 | 83 | |
AK074865.1 | - | 6 - 7 | 1189 | 83 | |
AK075014.1 | - | 6 - 7 | 1194 | 83 | |
BC033816.1 | - | 6 - 7 | 1173 | 83 |